Anti-SELE Rat Monoclonal Antibody (Biotin) [clone: 96409]

Supplier: R & D Systems
BAM5752
220014-638EA 728.86 USD
220014-638
Anti-SELE Rat Monoclonal Antibody (Biotin) [clone: 96409]
Antibodies
Type: Primary
Antigen: SELE
Clonality: Monoclonal
Clone: 96409
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR