Anti-CD27 Rat Monoclonal Antibody (Biotin) [clone: 137925]

Supplier: R & D Systems
BAM5741
220014-637EA 746.22 USD
220014-637
Anti-CD27 Rat Monoclonal Antibody (Biotin) [clone: 137925]
Antibodies
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: 137925
Conjugation: Biotin
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR