Anti-CD4 Rat Monoclonal Antibody (Biotin) [clone: GK1.5]

Supplier: R & D Systems
BAM554
220014-636EA 728.86 USD
220014-636
Anti-CD4 Rat Monoclonal Antibody (Biotin) [clone: GK1.5]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR