Anti-RPS6 Mouse Monoclonal Antibody (Biotin) [clone: 522731]

Supplier: R & D Systems
BAM5436
102879-142EA 698.49 USD
102879-142
Anti-RPS6 Mouse Monoclonal Antibody (Biotin) [clone: 522731]
Antibodies
Type: Primary
Antigen: RPS6
Clonality: Monoclonal
Clone: 522731
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR