Anti-NOTCH1 Mouse Monoclonal Antibody (Biotin) [clone: 527422]

Supplier: R & D Systems
BAM53171
220014-635EA 728.86 USD
220014-635
Anti-NOTCH1 Mouse Monoclonal Antibody (Biotin) [clone: 527422]
Antibodies
Type: Primary
Antigen: NOTCH1
Clonality: Monoclonal
Clone: 527422
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR