Anti-IL34 Mouse Monoclonal Antibody (Biotin) [clone: 578401]

Supplier: R & D Systems
BAM52652
102879-140EA 746.22 USD
102879-140
Anti-IL34 Mouse Monoclonal Antibody (Biotin) [clone: 578401]
Antibodies
Type: Primary
Antigen: IL34
Clonality: Monoclonal
Clone: 578401
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR