Anti-TNFA Mouse Monoclonal Antibody (Biotin) [clone: 569806]

Supplier: R & D Systems
BAM50351
102879-138EA 746.22 USD
102879-138
Anti-TNFA Mouse Monoclonal Antibody (Biotin) [clone: 569806]
Antibodies
Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: 569806
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR