Anti-CCL6 Rat Monoclonal Antibody (Biotin) [clone: 262008]

Supplier: R & D Systems
BAM4872
102878-976EA 746.22 USD
102878-976
Anti-CCL6 Rat Monoclonal Antibody (Biotin) [clone: 262008]
Antibodies
Type: Primary
Antigen: CCL6
Clonality: Monoclonal
Clone: 262008
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR