Anti-CD247 Rat Monoclonal Antibody (Biotin) [clone: 17A2]

Supplier: R & D Systems
BAM4841
220014-634EA 728.86 USD
220014-634
Anti-CD247 Rat Monoclonal Antibody (Biotin) [clone: 17A2]
Antibodies
Type: Primary
Antigen: CD247
Clonality: Monoclonal
Clone: 17A2
Conjugation: Biotin
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR