Anti-LAMP1 Mouse Monoclonal Antibody (Biotin) [clone: 508921]

Supplier: R & D Systems
BAM4800
102879-134EA 714.4 USD
102879-134
Anti-LAMP1 Mouse Monoclonal Antibody (Biotin) [clone: 508921]
Antibodies
Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: 508921
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR