Anti-IL7R Rat Monoclonal Antibody (Biotin) [clone: 132220]

Supplier: R & D Systems
BAM47741
220014-633EA 746.22 USD
220014-633
Anti-IL7R Rat Monoclonal Antibody (Biotin) [clone: 132220]
Antibodies
Type: Primary
Antigen: IL7R
Clonality: Monoclonal
Clone: 132220
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR