Anti-IL6ST Rat Monoclonal Antibody (Biotin) [clone: 125623]

Supplier: R & D Systems
BAM4681
220014-632EA 543.75 USD
220014-632
Anti-IL6ST Rat Monoclonal Antibody (Biotin) [clone: 125623]
Antibodies
Type: Primary
Antigen: IL6ST
Clonality: Monoclonal
Clone: 125623
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR