Anti-SHH Rat Monoclonal Antibody (Biotin) [clone: 171028]

Supplier: R & D Systems
BAM4642
220014-631EA 689.82 USD
220014-631
Anti-SHH Rat Monoclonal Antibody (Biotin) [clone: 171028]
Antibodies
Type: Primary
Antigen: SHH
Clonality: Monoclonal
Clone: 171028
Conjugation: Biotin
Epitope: N-Terminal
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR