Anti-CD40 Rat Monoclonal Antibody (Biotin) [clone: 1C10]

Supplier: R & D Systems
BAM440
220014-630EA 728.86 USD
220014-630
Anti-CD40 Rat Monoclonal Antibody (Biotin) [clone: 1C10]
Antibodies
Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 1C10
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR