Anti-CCL11 Rat Monoclonal Antibody (Biotin) [clone: 42285]

Supplier: R & D Systems
BAM420
220014-629EA 728.86 USD
220014-629
Anti-CCL11 Rat Monoclonal Antibody (Biotin) [clone: 42285]
Antibodies
Type: Primary
Antigen: CCL11
Clonality: Monoclonal
Clone: 42285
Conjugation: Biotin
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR