Anti-DMP1 Mouse Monoclonal Antibody (Biotin) [clone: 560722]

Supplier: R & D Systems
BAM41291
102879-130EA 728.86 USD
102879-130
Anti-DMP1 Mouse Monoclonal Antibody (Biotin) [clone: 560722]
Antibodies
Type: Primary
Antigen: DMP1
Clonality: Monoclonal
Clone: 560722
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR