Anti-LEP Mouse Monoclonal Antibody (Biotin) [clone: 44804]

Supplier: R & D Systems
BAM398
220014-628EA 769.35 USD
220014-628
Anti-LEP Mouse Monoclonal Antibody (Biotin) [clone: 44804]
Antibodies
Type: Primary
Antigen: LEP
Clonality: Monoclonal
Clone: 44804
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR