Anti-THBD Mouse Monoclonal Antibody (Biotin) [clone: 501727]

Supplier: R & D Systems
BAM39471
220014-627EA 728.86 USD
220014-627
Anti-THBD Mouse Monoclonal Antibody (Biotin) [clone: 501727]
Antibodies
Type: Primary
Antigen: THBD
Clonality: Monoclonal
Clone: 501727
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR