Anti-PECAM1 Mouse Monoclonal Antibody (Biotin) [clone: 9G11]

Supplier: R & D Systems
BAM3567
220014-625EA 723.08 USD
220014-625
Anti-PECAM1 Mouse Monoclonal Antibody (Biotin) [clone: 9G11]
Antibodies
Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 9G11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR