Anti-BMP7 Mouse Monoclonal Antibody (Biotin) [clone: 164324]

Supplier: R & D Systems
BAM354
220014-623EA 762.12 USD
220014-623
Anti-BMP7 Mouse Monoclonal Antibody (Biotin) [clone: 164324]
Antibodies
Type: Primary
Antigen: BMP7
Clonality: Monoclonal
Clone: 164324
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR