Anti-MST1 Mouse Monoclonal Antibody (Biotin) [clone: 68801]

Supplier: R & D Systems
BAM3521
220014-622EA 714.4 USD
220014-622
Anti-MST1 Mouse Monoclonal Antibody (Biotin) [clone: 68801]
Antibodies
Type: Primary
Antigen: MST1
Clonality: Monoclonal
Clone: 68801
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR