Anti-ERBB3 Mouse Monoclonal Antibody (Biotin) [clone: 66201]

Supplier: R & D Systems
BAM348
220014-620EA 728.86 USD
220014-620
Anti-ERBB3 Mouse Monoclonal Antibody (Biotin) [clone: 66201]
Antibodies
Type: Primary
Antigen: ERBB3
Clonality: Monoclonal
Clone: 66201
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR