Anti-RBP4 Mouse Monoclonal Antibody (Biotin) [clone: 393004]

Supplier: R & D Systems
BAM33782
102879-124EA 762.12 USD
102879-124
Anti-RBP4 Mouse Monoclonal Antibody (Biotin) [clone: 393004]
Antibodies
Type: Primary
Antigen: RBP4
Clonality: Monoclonal
Clone: 393004
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR