Anti-TEK Mouse Monoclonal Antibody (Biotin) [clone: 83704]

Supplier: R & D Systems
BAM3313
220014-617EA 746.22 USD
220014-617
Anti-TEK Mouse Monoclonal Antibody (Biotin) [clone: 83704]
Antibodies
Type: Primary
Antigen: TEK
Clonality: Monoclonal
Clone: 83704
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR