Anti-DAND5 Mouse Monoclonal Antibody (Biotin) [clone: 322206]

Supplier: R & D Systems
BAM3047
220014-614EA 728.86 USD
220014-614
Anti-DAND5 Mouse Monoclonal Antibody (Biotin) [clone: 322206]
Antibodies
Type: Primary
Antigen: DAND5
Clonality: Monoclonal
Clone: 322206
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR