Anti-THPO Mouse Monoclonal Antibody (Biotin) [clone: 34840]

Supplier: R & D Systems
BAM288
220014-612EA 714.4 USD
220014-612
Anti-THPO Mouse Monoclonal Antibody (Biotin) [clone: 34840]
Antibodies
Type: Primary
Antigen: THPO
Clonality: Monoclonal
Clone: 34840
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR