Anti-FIGF Mouse Monoclonal Antibody (Biotin) [clone: 78923]

Supplier: R & D Systems
BAM286
220014-611EA 746.22 USD
220014-611
Anti-FIGF Mouse Monoclonal Antibody (Biotin) [clone: 78923]
Antibodies
Type: Primary
Antigen Symbol: FIGF
Antigen Name: c-fos induced growth factor (vascular endothelial growth factor D)
Antigen Synonyms: VEGF-D, VEGFD
Clonality: Monoclonal
Clone: 78923
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR