Anti-IL24 Rat Monoclonal Antibody (Biotin) [clone: 303308]

Supplier: R & D Systems
BAM2786
220014-610EA 714.4 USD
220014-610
Anti-IL24 Rat Monoclonal Antibody (Biotin) [clone: 303308]
Antibodies
Type: Primary
Antigen: IL24
Clonality: Monoclonal
Clone: 303308
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR