Anti-TDGF1 Mouse Monoclonal Antibody (Biotin) [clone: 89627]

Supplier: R & D Systems
BAM2773
220014-609EA 746.22 USD
220014-609
Anti-TDGF1 Mouse Monoclonal Antibody (Biotin) [clone: 89627]
Antibodies
Type: Primary
Antigen: TDGF1
Clonality: Monoclonal
Clone: 89627
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR