Anti-CD200 Mouse Monoclonal Antibody (Biotin) [clone: 325523]

Supplier: R & D Systems
BAM27242
220014-608EA 746.22 USD
220014-608
Anti-CD200 Mouse Monoclonal Antibody (Biotin) [clone: 325523]
Antibodies
Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone: 325523
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR