Anti-REL Rat Monoclonal Antibody (Biotin) [clone: 290512]

Supplier: R & D Systems
BAM2699
220014-607EA 681.14 USD
220014-607
Anti-REL Rat Monoclonal Antibody (Biotin) [clone: 290512]
Antibodies
Type: Primary
Antigen: REL
Clonality: Monoclonal
Clone: 290512
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR