Anti-GRN Rat Monoclonal Antibody (Biotin) [clone: 333729]

Supplier: R & D Systems
BAM25572
102879-122EA 746.22 USD
102879-122
Anti-GRN Rat Monoclonal Antibody (Biotin) [clone: 333729]
Antibodies
Type: Primary
Antigen Symbol: GRN
Antigen Name: granulin
Antigen Synonyms: Pgrn, epithelin
Clonality: Monoclonal
Clone: 333729
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR