Anti-TGFB1 Mouse Monoclonal Antibody (Biotin) [clone: 27240]

Supplier: R & D Systems
BAM2462
220014-605EA 516 USD
220014-605
Anti-TGFB1 Mouse Monoclonal Antibody (Biotin) [clone: 27240]
Antibodies
Type: Primary
Antigen: TGFB1
Clonality: Monoclonal
Clone: 27240
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR