Anti-MOG Rat Monoclonal Antibody (Biotin) [clone: 296829]

Supplier: R & D Systems
BAM2439
220014-604EA 723.08 USD
220014-604
Anti-MOG Rat Monoclonal Antibody (Biotin) [clone: 296829]
Antibodies
Type: Primary
Antigen: MOG
Clonality: Monoclonal
Clone: 296829
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR