Anti-TGF-beta 3 Mouse Monoclonal Antibody (Biotin) [clone: 20724]

Supplier: R & D Systems
BAM243
220014-603EA 728.86 USD
220014-603
Anti-TGF-beta 3 Mouse Monoclonal Antibody (Biotin) [clone: 20724]
Antibodies
Type: Primary
Antigen Symbol: TGF-beta 3
Antigen Name:
Antigen Synonyms:
Clonality: Monoclonal
Clone: 20724
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR