Anti-FGF2 Mouse Monoclonal Antibody (Biotin) [clone: 10043]

Supplier: R & D Systems
BAM233
220014-601EA 769.35 USD
220014-601
Anti-FGF2 Mouse Monoclonal Antibody (Biotin) [clone: 10043]
Antibodies
Type: Primary
Antigen Symbol: FGF2
Antigen Name: fibroblast growth factor 2 (basic)
Antigen Synonyms: BFGF, FGF-2, FGFB, HBGF-2
Clonality: Monoclonal
Clone: 10043
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR