Anti-RARRES2 Rat Monoclonal Antibody (Biotin) [clone: 372402]

Supplier: R & D Systems
BAM2325
220014-600EA 746.22 USD
220014-600
Anti-RARRES2 Rat Monoclonal Antibody (Biotin) [clone: 372402]
Antibodies
Type: Primary
Antigen: RARRES2
Clonality: Monoclonal
Clone: 372402
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR