Anti-CXCL8 Rat Monoclonal Antibody (Biotin) [clone: 344610]

Supplier: R & D Systems
BAM22771
220014-598EA 728.86 USD
220014-598
Anti-CXCL8 Rat Monoclonal Antibody (Biotin) [clone: 344610]
Antibodies
Type: Primary
Antigen: CXCL8
Clonality: Monoclonal
Clone: 344610
Conjugation: Biotin
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR