Anti-CTSB Mouse Monoclonal Antibody (Biotin) [clone: 155709]

Supplier: R & D Systems
BAM21761
220014-596EA 714.4 USD
220014-596
Anti-CTSB Mouse Monoclonal Antibody (Biotin) [clone: 155709]
Antibodies
Type: Primary
Antigen: CTSB
Clonality: Monoclonal
Clone: 155709
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR