Anti-CSF2 Mouse Monoclonal Antibody (Biotin) [clone: 3209]

Supplier: R & D Systems
BAM215
220014-595EA 769.35 USD
220014-595
Anti-CSF2 Mouse Monoclonal Antibody (Biotin) [clone: 3209]
Antibodies
Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: 3209
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR