Anti-WFIKKN1 Mouse Monoclonal Antibody (Biotin) [clone: 289839]

Supplier: R & D Systems
BAM21362
220014-594EA 728.86 USD
220014-594
Anti-WFIKKN1 Mouse Monoclonal Antibody (Biotin) [clone: 289839]
Antibodies
Type: Primary
Antigen: WFIKKN1
Clonality: Monoclonal
Clone: 289839
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR