Anti-TNFRSF6B Mouse Monoclonal Antibody (Biotin) [clone: 229812]

Supplier: R & D Systems
BAM2135
220014-593EA 746.22 USD
220014-593
Anti-TNFRSF6B Mouse Monoclonal Antibody (Biotin) [clone: 229812]
Antibodies
Type: Primary
Antigen: TNFRSF6B
Clonality: Monoclonal
Clone: 229812
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR