Anti-VIM Rat Monoclonal Antibody (Biotin) [clone: 280618]

Supplier: R & D Systems
BAM2105
102879-120EA 594.37 USD
102879-120
Anti-VIM Rat Monoclonal Antibody (Biotin) [clone: 280618]
Antibodies
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 280618
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR