Anti-PRSS22 Rat Monoclonal Antibody (Biotin) [clone: 246703]

Supplier: R & D Systems
BAM20591
220014-592EA 728.86 USD
220014-592
Anti-PRSS22 Rat Monoclonal Antibody (Biotin) [clone: 246703]
Antibodies
Type: Primary
Antigen: PRSS22
Clonality: Monoclonal
Clone: 246703
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR