Anti-C5 Mouse Monoclonal Antibody (Biotin) [clone: 295009]

Supplier: R & D Systems
BAM20371
220014-591EA 762.12 USD
220014-591
Anti-C5 Mouse Monoclonal Antibody (Biotin) [clone: 295009]
Antibodies
Type: Primary
Antigen: C5
Clonality: Monoclonal
Clone: 295009
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR