Anti-FZD7 Rat Monoclonal Antibody (Biotin) [clone: 151143]

Supplier: R & D Systems
BAM1981
220014-590EA 689.82 USD
220014-590
Anti-FZD7 Rat Monoclonal Antibody (Biotin) [clone: 151143]
Antibodies
Type: Primary
Antigen Symbol: FZD7
Antigen Name: frizzled homolog 7 (Drosophila)
Antigen Synonyms: Fz7
Clonality: Monoclonal
Clone: 151143
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR