Anti-FZD4 Rat Monoclonal Antibody (Biotin) [clone: 145901]

Supplier: R & D Systems
BAM194
220014-587EA 543.75 USD
220014-587
Anti-FZD4 Rat Monoclonal Antibody (Biotin) [clone: 145901]
Antibodies
Type: Primary
Antigen Symbol: FZD4
Antigen Name: frizzled homolog 4 (Drosophila)
Antigen Synonyms: Fz4
Clonality: Monoclonal
Clone: 145901
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR