Anti-FRZB Mouse Monoclonal Antibody (Biotin) [clone: 133912]

Supplier: R & D Systems
BAM192
220014-586EA 746.22 USD
220014-586
Anti-FRZB Mouse Monoclonal Antibody (Biotin) [clone: 133912]
Antibodies
Type: Primary
Antigen Symbol: FRZB
Antigen Name: frizzled-related protein
Antigen Synonyms: FRE, FRITZ, FRP-3, FRZB-1, FRZB-PEN, FRZB1, FZRB, OS1, SFRP3, SRFP3, hFIZ
Clonality: Monoclonal
Clone: 133912
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR