Anti-SLAMF8 Mouse Monoclonal Antibody (Biotin) [clone: 250008]

Supplier: R & D Systems
BAM19074
220014-585EA 728.86 USD
220014-585
Anti-SLAMF8 Mouse Monoclonal Antibody (Biotin) [clone: 250008]
Antibodies
Type: Primary
Antigen: SLAMF8
Clonality: Monoclonal
Clone: 250008
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR