Anti-FCGR2B Mouse Monoclonal Antibody (Biotin) [clone: 190703]

Supplier: R & D Systems
BAM1875
220014-584EA 746.22 USD
220014-584
Anti-FCGR2B Mouse Monoclonal Antibody (Biotin) [clone: 190703]
Antibodies
Type: Primary
Antigen Symbol: FCGR2B
Antigen Name: Fc fragment of IgG, low affinity IIb, receptor (CD32)
Antigen Synonyms: CD32, CD32B, FCG2, FCGR2, IGFR2
Clonality: Monoclonal
Clone: 190703
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR